Learn More
Vidaro™ Open Face Polyester Cleanroom Hoods
Catalog No. 19999865 Shop All Vidaro Corporation Products
Change view
Click to view available options
Color:
Light Blue
White
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-999-865 Light Blue X-Large
19-999-867 White X-Small
19-999-868 White Small
19-999-869 White Medium
19-999-870 White Large
19-999-871 White X-Large
19-999-872 White 2X-Large
19-999-861 Light Blue X-Small
19-999-862 Light Blue Small
19-999-863 Light Blue Medium
19-999-864 Light Blue Large
19-999-866 Light Blue 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Catalog No. 19-999-865 Supplier Vidaro™ Supplier No. H3311/XL
This item is not returnable. View return policy
Cleanroom hoods feature a stainless steel front-snap closure and are ideal for critical environments. Choose blue or white color in a variety of sizes.
  • Open face polyester cleanroom hoods
  • Sizes range from x-small to 2X-large
  • Polyester material
  • Two snap closure on each cheek
  • Stainless steel front-snap closure
  • Adjustable back snap at neck
Specifications
Product Type Open Face
Size X-Large
Material Polyester
Color Light Blue
Closure Type Snap Front
Disposable Single-use
Model H-3311
Quantity 1 Ea.
Includes Two snaps on each cheek, Stainless Steel front-snap closure, adjustable back snap at neck