Learn More
Vidaro™ Polyester Cleanroom Frocks
Catalog No. 19999780 Shop All Vidaro Corporation Products
Change view
Click to view available options
Color:
Blue
White
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-999-780 Blue
19-999-772 White
19-999-756 White
19-999-773 White
19-999-757 White
19-999-768 White
19-999-770 White
19-999-764 Blue
19-999-781 Blue
19-999-763 Blue
19-999-765 Blue
19-999-762 Blue
19-999-760 Blue
19-999-761 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Catalog No. 19-999-780 Supplier Vidaro™ Supplier No. SM3311/3XL
This item is not returnable. View return policy
Comfortable and durable
  • Made of 3 oz. (85g) per square yard, 150 denier, limited-linting 100% polyester fabric
  • Major seams are serged and double-needle (felled) stitched for strength and cleanliness
  • Military collar with color-coded hanger loop for size identification
  • Stainless-steel snap-front closure allows for adjustment at neck
  • Long raglan sleeves with snap or knit cuff
  • Available in white or blue
Specifications
Material Polyester
Color Blue
Disposable Reusable
Product Type Controlled Environments Frock
Seam Type Serged, Double-needle Stitched