Learn More
Vidaro™ Vi-Gard™ I Static-Control Lab Coats
Catalog No. 19999633 Shop All Vidaro Corporation Products
Change view
Click to view available options
Color:
Blue
Royal Blue
White
Material:
Polyester
Polyester, Cotton, Carbon-modified Nylon
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Material Size
19-999-633 Royal Blue Polyester, Cotton, Carbon-modified Nylon Medium
19-999-620 White Polyester, Cotton, Carbon-modified Nylon Large
19-999-619 White Polyester, Cotton, Carbon-modified Nylon Medium
19-999-618 White Polyester, Cotton, Carbon-modified Nylon Small
19-999-629 Blue Polyester, Cotton, Carbon-modified Nylon 2X-Large
19-999-630 Blue Polyester, Cotton, Carbon-modified Nylon 3X-Large
19-999-628 Blue Polyester, Cotton, Carbon-modified Nylon X-Large
19-999-636 Royal Blue Polyester, Cotton, Carbon-modified Nylon 2X-Large
19-999-637 Royal Blue Polyester, Cotton, Carbon-modified Nylon 3X-Large
19-999-632 Royal Blue Polyester Small
19-999-635 Royal Blue Polyester, Cotton, Carbon-modified Nylon X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 19-999-633 Supplier Vidaro™ Supplier No. L63176/M
This item is not returnable. View return policy
Static control is designed into this state-of-the-art ESD fabric
  • Blends 65% polyester, 34% cotton, and 1% carbon-modified nylon yarn for durable garments with permanent static control
  • Cool and comfortable enough to wear over regular clothing
  • Effectively reduce electrostatic charges to safe levels and eliminate arcing from workers to sensitive electronic products
  • Coats are 38 in. long (97cm)
  • Long, hemmed sleeves, left breast pocket with pencil slot, two lower front pockets, laydown collar, and side slits for access to inner garments
  • Front closure features five stainless-steel snaps
Specifications
Product Type Controlled Environments Lab Coat
Size Medium
Material Polyester, Cotton, Carbon-modified Nylon
Color Royal Blue
Closure Type Snaps
Cuff Style Hemmed
Disposable Reusable
Collar Type Lay Down
Model L-6317-6
Size (English) Chest 38 to 40 in.
Length (English) 38 in.
Length (Metric) 96.52 cm