Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VIGR/GPR126 Antibody (1044430), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB10577C
This item is not returnable.
View return policy
Description
VIGR/GPR126 Monoclonal antibody specifically detects VIGR/GPR126 in Human samples. It is validated for Flow CytometrySpecifications
VIGR/GPR126 | |
Monoclonal | |
PerCP | |
APG1, Developmentally regulated G-protein-coupled receptor, DREG, FLJ14937, G protein-coupled receptor 126, G-protein coupled receptor 126, HBV PreS1-transactivated protein 2, Vascular inducible G protein-coupled receptor, vascular-inducible G protein-coupled receptor, VIGRPS1TP2 | |
Human embryonic kidney cell HEK293-derived human VIGR/GPR126 protein, Cys38-Lys437, Accession # AAH75798.1 | |
0.1 mL | |
GPCR | |
57211 | |
Store at 4°C in the dark. | |
IgG2a |
Flow Cytometry | |
1044430 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction