Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human VIGR/GPR126 Alexa Fluor™ 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB10577V100UG
This item is not returnable.
View return policy
Description
VIGR/GPR126 Monoclonal antibody specifically detects VIGR/GPR126 in Human samples. It is validated for Flow CytometrySpecifications
VIGR/GPR126 | |
Monoclonal | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
APG1, Developmentally regulated G-protein-coupled receptor, DREG, FLJ14937, G protein-coupled receptor 126, Gm222, GPR126, G-protein coupled receptor 126, HBV PreS1-transactivated protein 2, PS1TP2, Vascular inducible G protein-coupled receptor, vascular-inducible G protein-coupled receptor, VIGRPS1TP2 | |
Human embryonic kidney cell HEK293-derived human VIGR/GPR126 protein, Cys38-Lys437, Accession # AAH75798.1 | |
100 μg | |
Primary | |
Reconstitution default not known | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG2a |
Flow Cytometry | |
1044430 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
AAH75798.1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
57211 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction