Learn More
United Scientific Supplies Vigreux Distilling Columns, 24/40 Joint
SDPSCI_ED
Catalog No. S43794
Click to view available options
Length (Metric):
200 mm
300 mm
400 mm
500 mm
Allows for a dramatic increase in surface area per unit length
  • Vigreux columns have a modified sample tube to include an abundance of downward-pointing indentations
  • This design dramatically increases the surface area per unit length of the condenser
  • Includes a 24/40 connector
Specifications
For Use With (Equipment) Air Condenser
Material Borosilicate Glass
Type Vigreux Distilling Column
For Use With (Application) Distillation
Connection Type 24/40
Length (Metric) 400 mm
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.