Learn More
Vikan™ 3 Gallon Bucket
Catalog No. 17227518 Shop All Remco Products
Change view
Click to view available options
Color:
Blue
Green
Orange
Pink
Purple
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-227-518 Pink
17-227-521 Orange
17-227-519 Green
17-227-520 Blue
17-227-522 Purple
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-227-518 Supplier Vikan™ Supplier No. 56861
Add to Cart
Add to Cart
Vikan™ 3 Gallon Bucket is calibrated for a variety of measuring units and is suitable for cleaning or storage of ingredients and food stuff.
  • Ideal for transporting cleaning chemicals as well as hot or cold ingredients
  • Drip-free spout
  • Hand grip that is raised from the base
  • Sturdy stainless steel handle
  • Flat side prevents spillage and stores easily using the coordinating wall bracket
  • Updated grips along the top of the bucket allow for a lid to snap in place
  • Color-coded
Specifications
Product Type Bucket
Material Polypropylene, Stainless Steel
Autoclavable Not Autoclavable
Capacity (English) 3 gal.
Certifications/Compliance FDA Compliant
Color Pink
Dimensions 12.81 x 12.88 in.
Height (English) 12.5 in.
Quantity 1 Ea.
Sterility Non-sterile