Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vikan™ Color-Coded Wall Bracket Replacement Hooks

Catalog No. 17227013 Shop All Remco Products
Click to view available options
Color:
Black
Blue
Green
Orange
Pink
Purple
Red
White
Yellow

Vikan™ Color-Coded Wall Bracket Replacement Hooks combine with a wall bracket system to allow for total customization of your storage systems.

  • Contains 4 hooks and 8 pins
  • For use with wall brackets 1017 and 1018
  • Allows total customization of the wall bracket system to fit any color-coded program
  • Extra hooks can be added to hold more hand tools such as brushes, scrapers, and squeegees
  • Hold more smaller hand tools with these easy-to-install hooks

Specifications

Product Type Hooks
Color Red
Material Polypropylene
For Use With (Application) Customizing wall bracket storage systems
Certifications/Compliance FDA Compliant
Quantity 1
Dimensions (L x W x H) 3 x 0.8 x 2.4 in.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.