Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vikan™ Extra-Stiff 11 in. Waterfed Deck Scrub

Catalog No. 17227776 Shop All Remco Products
Change view
Click to view available options
Color:
Blue
Green
Orange
Red
White
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-227-776 White
17-227-775 Red
17-227-778 Orange
17-227-777 Yellow
17-227-773 Green
17-227-774 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17-227-776 Supplier Vikan™ Supplier No. 70415
Add to Cart
Add to Cart

Vikan™ Extra-Stiff 11 in. Waterfed Deck Scrub is an extremely stiff, short-bristled scrubbing broom that is ideal for cleaning in narrow spaces.

  • Broom can withstand heavy downward force from the user
  • Small size makes it ideal for cleaning in narrow spaces
  • Color-coded

Specifications

Product Type Scrub Brush
Color-coded Yes
Handle Material Polypropylene
Sterility Non-sterile
Certifications/Compliance FDA Compliant
Color White
Description Thread type Euro Hygienic
Length (English) 10.75 in.
Quantity 1 Ea.
Size Large
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.