Learn More
Vikan™ Extra Stiff Detail Brush
Catalog No. 17227330 Shop All Remco Products
Change view
Click to view available options
Color:
Blue
Green
Red
White
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-227-330 Yellow
17-227-329 Red
17-227-327 Green
17-227-328 Blue
19-830-225 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-227-330 Supplier Vikan™ Supplier No. 44016
Add to Cart
Add to Cart
Vikan™ Extra Stiff Detail Brush is designed with a smooth, ergonomic handle for cleaning in and around the gaskets and joints of machinery.
  • Small and compact design
  • Suitable for cleaning in and around gaskets, rubber strips, and joints on machinery
  • Can also be used as a grout brush and for cleaning equipment link conveyor belts
  • Color-coded
Specifications
Product Type Cleaning Brush
Color-coded Yes
Handle Material Polypropylene
Sterility Non-sterile
Certifications/Compliance FDA Compliant
Color Yellow
Length (English) 7.9 in.
Quantity 1 Ea.
Size Small