Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vikan™ Lid for 3 Gallon Bucket

Catalog No. 17227529 Shop All Remco Products
Change view
Click to view available options
Color:
Green
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-227-529 -
17-227-526 -
17-227-528 -
17-227-524 Green
17-227-525 -
17-227-530 -
17-227-523 -
17-227-527 -
17-227-531 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 17-227-529 Supplier Vikan™ Supplier No. 56877
Add to Cart
Add to Cart

Vikan™ Lid for 3 Gallon Bucket has a clip system for keeping the lid in place when in use.

  • Ideal for preventing spillage of materials
  • It has a clip system for securing the lid tightly
  • For use with 3 Gallon Bucket
  • Color-coded

Specifications

Autoclavable Not Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.