Description
- Allows you to reach into parts of equipment that are hard to access
- Angled bristles are stiff, helping to scrub at a distance
- Color-coded
Specifications
Specifications
| Product Type | Scrub Brush |
| Color-coded | Yes |
| Handle Material | Polypropylene |
| Sterility | Non-sterile |
| Certifications/Compliance | FDA Compliant |
| Color | White |
| Length (English) | 15.75 in. |
| Quantity | 1 Ea. |
| Size | Medium |