Learn More
Vikan™ Meadium/Stiff One-Piece Pipe Brush
Catalog No. 17227452 Shop All Remco Products
Change view
Click to view available options
Color:
Black
Blue
Green
Orange
Purple
Red
White
Yellow
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-227-452 Blue
17-227-453 Red
17-227-456 Orange
17-227-455 Yellow
17-227-451 Green
17-227-457 Purple
17-227-454 White
17-227-458 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-227-452 Supplier Vikan™ Supplier No. 5381903
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
Vikan™ Meadium/Stiff One-Piece Pipe Brush is manufactured with no joints between the handle and brush head.
  • Features bristles along the sides and in the front
  • It is used in areas where a wider tube cleaning action is required or where the equipment has an endpoint that cannot be reached with a hand brush
  • Works well as a drain brush
  • Color-coded
Specifications
Product Type Pipe Brush
Color-coded Yes
Handle Material Polypropylene
Sterility Non-sterile
Certifications/Compliance FDA Compliant
Color Blue
Diameter (English) Bristle 3.5 in.
Length (English) 16.9 in.
Quantity 1 Ea.
Size Medium