Description
- Allows easy cleaning between and around pipes
- Curved design
- The hand grip utilizes a threaded recess on the end that allows another handle to be connected to the brush
- When connected to a Vikan™ handle, the extended hook brush allows comfortable cleaning of pipes in high or difficult to reach areas
Specifications
Specifications
| Product Type | Pipe Brush |
| Color-coded | Yes |
| Handle Material | Stainless Steel |
| Sterility | Non-sterile |
| Certifications/Compliance | FDA Compliant |
| Color | Black |
| Length (English) | 15.5 in. |
| Quantity | 1 Ea. |
| Size | Medium |