Learn More
Vikan™ Medium High-Low Brush
Catalog No. 17227794 Shop All Remco Products
Change view
Click to view available options
Color:
Black
Blue
Green
Orange
Pink
Red
White
Yellow
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-227-794 Yellow
17-227-792 Red
17-227-795 Orange
17-227-790 Green
17-227-791 Blue
17-227-789 Pink
17-227-793 White
17-227-796 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-227-794 Supplier Vikan™ Supplier No. 70476
Add to Cart
Add to Cart
Vikan™ Medium High-Low Brush has bristles strong enough to be used on equipment and in the areas where the wall meets the floor.
  • Angled shape of this scrubbing broom was designed to clean the area where the wall meets the floor
  • Effective for cleaning the floor
  • Bristles are gentle enough to be used on equipment
  • Ideal choice in meat processing facilities, canneries, mills, wineries and breweries, and generally any moist environment that creates difficult cleaning challenges
  • Color-coded
Specifications
Product Type Cleaning Brush
Color-coded Yes
Handle Material Polypropylene
Sterility Non-sterile
Certifications/Compliance FDA Compliant
Color Yellow
Description Thread type Euro Hygienic
Length (English) 10.4 in.
Quantity 1 Ea.
Size Large