Description
- Ideal for sweeping damp particles from conveyor belts, food preparation surfaces, tables, and equipment in high risk areas
- The placement of the Bristle Security Units enables efficient sweeping of damp and wet particles
- Color-coded
Specifications
Specifications
| Product Type | Bench Brush |
| Color-coded | Yes |
| Handle Material | Polypropylene |
| Sterility | Non-sterile |
| Certifications/Compliance | FDA Compliant |
| Color | Green |
| Length (English) | 13 in. |
| Quantity | 1 Ea. |
| Size | Medium |