Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vikan™ Soft 11 in. Waterfed Deck Scrub

Catalog No. 17227782 Shop All Remco Products
Change view
Click to view available options
Color:
Blue
Green
Red
White
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-227-782 White
17-227-781 Red
17-227-783 Yellow
17-227-779 Green
17-227-780 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-227-782 Supplier Vikan™ Supplier No. 70435
Add to Cart
Add to Cart

Vikan™ Soft 11 in. Waterfed Deck Scrub can be used in a waterfed system or with a standard Euro-thread handle.

  • Scrub has effective water distribution
  • Ideal for use in heavily soiled areas such as meat processing plants
  • Color-coded

Specifications

Product Type Scrub Brush
Color-coded Yes
Handle Material Polypropylene
Sterility Non-sterile
Certifications/Compliance FDA Compliant
Color White
Description Thread type Euro Hygienic
Length (English) 10.9 in.
Quantity 1 Ea.
Size Large
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.