Learn More
Vikan™ Soft Bench Brush
Catalog No. 17227351 Shop All Remco Products
Change view
Click to view available options
Color:
Black
Blue
Green
Orange
Pink
Purple
Red
White
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-227-351 Green
17-227-353 Red
17-227-356 Orange
17-227-355 Yellow
17-227-352 Blue
17-227-357 Purple
17-227-350 Pink
17-227-354 White
17-227-358 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 17-227-351 Supplier Vikan™ Supplier No. 45872
Add to Cart
Add to Cart
Vikan™ Soft Bench Brush is a dusting brush with a smooth, ergonomically designed handle.
  • Smooth, ergonomically designed handle
  • The soft bristles are designed for brushing dry, fine particles, such as dust, flour, icing sugar
  • Can also be used as a dustpan brush on floors
  • Color-coded
Specifications
Product Type Bench Brush
Color-coded Yes
Handle Material Polypropylene
Sterility Non-sterile
Certifications/Compliance FDA Compliant
Color Green
Length (English) 13 in.
Quantity 1 Ea.
Size Medium