Learn More
Vikan™ Soft UST Bench Brush
Catalog No. 17227334 Shop All Remco Products
Change view
Click to view available options
Color:
Blue
Green
Orange
Pink
Purple
Red
White
Yellow
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-227-334 Red
17-227-337 Orange
17-227-336 Yellow
17-227-332 Green
17-227-333 Blue
17-227-338 Purple
17-227-331 Pink
17-227-335 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-227-334 Supplier Vikan™ Supplier No. 45814
Only null left
Add to Cart
Add to Cart
Vikan™ Soft UST Bench Brush is ideal for application that include flour and other fine ingredients.
  • Suitable for sweeping fine dry particles from conveyor belts, food preparation surfaces, tables, and equipment in high risk areas
  • The placement of the Bristle Security Units enables efficient sweeping of fine dry particles
  • Brush is easy to clean and hygienically designed
  • Color- coded
Specifications
Product Type Bench Brush
Color-coded Yes
Handle Material Polypropylene
Sterility Non-sterile
Certifications/Compliance FDA Compliant
Color Red
Length (English) 13 in.
Quantity 1 Ea.
Size Medium