Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vikan™ Stiff Paddle Scraper

Catalog No. p-7162003 Shop All Remco Products
Click to view available options
Color Code:
Blue
Green
Red
White
Yellow

Stiff paddle scraper is used to remove remaining food stuff from containers prior to the cleaning procedure.

  • Intended for scraping substances from the sides of containers
  • Ideal tool for prepping prior to the cleaning procedure
  • Euro-thread connection allows for pairing with any length or type of Vikan™ handle
  • Can be used on everything from deep containers to simple mixing bowls
  • Rigid construction is useful when firm pressure needs to be applied into deep containers
  • Color-coded

Specifications

Product Type Scraper
Color-coded Yes
Color Code Red
Certifications/Compliance FDA Compliant
Color Red
Material Nylon
Size Medium
Description Thread type Euro Hygienic
Dimensions (L x W) 8.75 x 4.3 in.
Quantity 1 Ea.
Includes Thread type Euro Hygienic
Weight (English) 0.32 lb.
No. per Pack 1 Ea.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.