Learn More
Vikan™ Stiff Round Hand Brush
Catalog No. 17227226 Shop All Remco Products
Change view
Click to view available options
Color:
Blue
Green
Red
White
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-227-226 Yellow
17-227-225 Red
19-830-218 Green
17-227-224 Blue
19-830-213 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-227-226 Supplier Vikan™ Supplier No. 38856
Add to Cart
Add to Cart
Vikan™ Stiff Round Hand Brush features a raised grip and stiff bristles that are ideal for cleaning tough messes while being ergonomically friendly.
  • Raised grip that helps keep hands from touching surfaces and chemicals
  • Designed to allow more pressure for better cleaning
  • Ideal for cleaning inside machines with small entry points
  • Stiff bristles are best used with large particles or to clean up tough messes
  • Color-coded
Specifications
Product Type Round Brush
Color-coded Yes
Handle Material Polypropylene
Sterility Non-sterile
Certifications/Compliance FDA Compliant
Color Yellow
Length (English) 5.1 in.
Quantity 1 Ea.
Width (English) Bristle 3.75 in.