Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VIL1 Rabbit anti-Human, Unconjugated, Clone: VIL1/2310R, Abnova™

Catalog No. 89339667 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-667 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-667 Supplier Abnova Corporation Supplier No. RAB00652
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

VIL1 Monoclonal antibody specifically detects VIL1 in Human samples. It is validated for IHC-P.

Specifications

Antigen VIL1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone VIL1/2310R
Conjugate Unconjugated
Description villin 1
Gene VIL1
Gene Alias D2S1471, VIL
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 179-311 of human VIL1
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 7429
Target Species Human
Content And Storage Store at -20 to -80°C
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.