Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VILIP 3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | VILIP 3 |
---|---|
Concentration | 0.28 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-6771
![]() ![]() |
Proteintech Group Inc
109891AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Hippocalcin-like protein 1, may be involved in the calcium-dependent regulation of rhodopsin phosphorylation.Specifications
VILIP 3 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P37235, P62748 | |
3241, 53602 | |
VILIP 3 Fusion Protein Ag1449 | |
Primary | |
-20°C | |
HPCAL1 |
0.28 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
BDR1, Calcium binding protein BDR 1, hippocalcin like 1, Hippocalcin like protein 1, HLP2, HPCA, HPCAL1, VILIP 3, Visinin like protein 3 | |
HPCAL1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title