Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Villin 1 Antibody (VIL1/2376), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308928AF488
Description
Villin 1 Monoclonal antibody specifically detects Villin 1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Villin 1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| D2S1471, VIL, villin 1, villin-1 | |
| A recombinant fragment (around aa 179-311) of human Villin 1 protein (exact sequence is proprietary) (Uniprot: P09327) | |
| 100 μg | |
| Cancer, Cell Biology, Cellular Markers | |
| 7429 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| VIL1/2376 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction