Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Villin 1 Antibody (VIL1/2376), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308928AF594
Description
Villin 1 Monoclonal antibody specifically detects Villin 1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Villin 1 | |
Monoclonal | |
Alexa Fluor 594 | |
D2S1471, VIL, villin 1, villin-1 | |
A recombinant fragment (around aa 179-311) of human Villin 1 protein (exact sequence is proprietary) (Uniprot: P09327) | |
100 μg | |
Cancer, Cell Biology, Cellular Markers | |
7429 | |
Store at 4°C in the dark. | |
IgG1 κ |
Immunohistochemistry (Paraffin) | |
VIL1/2376 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction