Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Villin 1 Antibody (VIL1/1314), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$268.00 - $524.00

Specifications

Antigen Villin 1
Clone VIL1/1314
Concentration 0.2 mg/mL
Dilution Western Blot 1 - 2 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.25 - 0.5 ug/ml
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB25319520
SDP
View Documents
Novus Biologicals
NBP25319520UG
20 μg
Each for $268.00
Only null left
Add to Cart
 
NB253195100
SDP
View Documents
Novus Biologicals
NBP253195100UG
100 μg
Each for $524.00
Only null left
Add to Cart
 
NB253195200
SDP
View Documents
Novus Biologicals
NBP253195200UG
200 μg N/A N/A N/A
Description

Description

Villin 1 Monoclonal specifically detects Villin 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Villin 1
0.2 mg/mL
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Cancer, Cytoskeleton Markers
10mM PBS with 0.05% BSA with 0.05% Sodium Azide
7429
Recombinant human Villin fragment of 133 amino acid residues (aa179-311) (exact sequence is proprietary)
Primary
Store at 4C.
VIL1/1314
Western Blot 1 - 2 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.25 - 0.5 ug/ml
Monoclonal
Purified
RUO
Human
D2S1471, VIL, villin 1, villin-1
VIL1
IgG1 κ
Protein A or G purified
93 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.