Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Villin 1 Rabbit anti-Human, Clone: VIL1/4107R, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP30719020UG
Description
Villin 1 Monoclonal specifically detects Villin 1 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
Villin 1 | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
D2S1471, VIL, villin 1, villin-1 | |
Recombinant fragment (around aa600-700) of human Villin 1 protein (exact sequence is proprietary) | |
20 μg | |
Cancer, Cell Biology, Cellular Markers | |
7429 | |
Store at 4°C. | |
IgG |
Immunohistochemistry (Paraffin) | |
VIL1/4107R | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction