Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Villin Monoclonal Antibody (CWWB1)
GREENER_CHOICE

Catalog No. PIMA512227
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA512227 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA512227 Supplier Invitrogen™ Supplier No. MA512227
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-12227 targets Villin in IHC (P) applications and shows reactivity with Human samples. The MA5-12227 immunogen is human Villin protein.

Villin is part of cytoskeleton associated with the microvillar actin core bundle of gastrointestinal and renal brush border. It can interact with actin in a Ca2+ and phosphoinositide-regulated manner. Villin is a very specific marker for adenocarcinomas of the gastrointestinal tract and the pancreas. It is also expressed in some Merkel cell tumor and adenocarcinoma of the lung, prostate, ovarian and kidney.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Villin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CWWB1
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene VIL1
Gene Accession No. P09327, Q62468
Gene Alias D2S1471; OTTHUMP00000207159; Vil; Vil1; villin 1; villin-1
Gene Symbols VIL1
Host Species Mouse
Immunogen Human Villin protein.
Purification Method Protein G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22349, 7429
Target Species Human, Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.