Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Villin Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578222
Description
Positive Control: HepG2, mouse kidney Predicted Reactivity: Rat (87%), Bovine (87%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Villin is part of cytoskeleton associated with the microvillar actin core bundle of gastrointestinal and renal brush border. It can interact with actin in a Ca2+ and phosphoinositide-regulated manner. Villin is a very specific marker for adenocarcinomas of the gastrointestinal tract and the pancreas. It is also expressed in some Merkel cell tumor and adenocarcinoma of the lung, prostate, ovarian and kidney.
Specifications
Villin | |
Polyclonal | |
Unconjugated | |
VIL1 | |
D2S1471; OTTHUMP00000207159; Vil; Vil1; villin 1; villin-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22349, 7429 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
P09327, Q62468 | |
VIL1 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of Human Villin. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction