Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Villin Recombinant Rabbit Monoclonal Antibody (A2)
GREENER_CHOICE

Catalog No. FEMA550679
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550679 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550679 Supplier Invitrogen™ Supplier No. MA550679
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive control: HT-29 cell lysates, human kidney tissue, human small intestine tissue. Predicted band size: 93 kDa Subcellular Location: Cytoskeleton, lamellipodium, ruffle, microvillus, filopodium tip, filopodium.

Villin is part of cytoskeleton associated with the microvillar actin core bundle of gastrointestinal and renal brush border. It can interact with actin in a Ca2+ and phosphoinositide-regulated manner. Villin is a very specific marker for adenocarcinomas of the gastrointestinal tract and the pancreas. It is also expressed in some Merkel cell tumor and adenocarcinoma of the lung, prostate, ovarian and kidney.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Villin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone A2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene VIL1
Gene Accession No. P09327
Gene Alias D2S1471; OTTHUMP00000207159; Vil; Vil1; villin 1; villin-1
Gene Symbols VIL1
Host Species Rabbit
Immunogen Recombinant protein within human Villin aa 600-827.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7429
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.