Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VIM Mouse anti-Human, Biotin, Clone: OTI7B10, liquid, TrueMAB™
SDP

Catalog No. 501677860
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-7860 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-7860 Supplier OriGene Supplier No. TA700390
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Vimentin is a member of the intermediate filament family. Intermediate filamentents, along with microtubules and actin microfilaments, make up the cytoskeleton. The protein is responsible for maintaining cell shape, integrity of the cytoplasm, and stabilizing cytoskeletal interactions. It is also involved in the immune response, and controls the transport of low-density lipoprotein (LDL)-derived cholesterol from a lysosome to the site of esterification. It functions as an organizer of a number of critical proteins involved in attachment, migration, and cell signaling. Mutations in this gene causes a domit, pulverulent cataract.

Specifications

Antigen VIM
Applications ELISA
Classification Monoclonal
Clone OTI7B10
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation PBS with 10mg/ml BSA, 50% glycerol and 0.05% sodium azide
Gene VIM
Gene Accession No. P08670
Gene Alias vimentin
Gene Symbols VIM
Host Species Mouse
Immunogen Full length human recombit protein of human VIM produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7431
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.