Learn More
VIM Mouse anti-Human, Clone: OTI2C4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF801310

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Vimentin is a member of the intermediate filament family. Intermediate filamentents, along with microtubules and actin microfilaments, make up the cytoskeleton. The protein is responsible for maintaining cell shape, integrity of the cytoplasm, and stabilizing cytoskeletal interactions. It is also involved in the immune response, and controls the transport of low-density lipoprotein (LDL)-derived cholesterol from a lysosome to the site of esterification. It functions as an organizer of a number of critical proteins involved in attachment, migration, and cell signaling. Mutations in this gene causes a domit, pulverulent cataract.Specifications
VIM | |
Monoclonal | |
Unconjugated | |
VIM | |
vimentin | |
Mouse | |
Affinity Chromatography | |
RUO | |
7431 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2C4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P08670 | |
VIM | |
Full length human recombit protein of human VIM produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.