Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VIM Rabbit anti-Human, Polyclonal, Abnova™

Catalog No. 89035668 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
40 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-668 40 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-668 Supplier Abnova Corporation Supplier No. PAB30173

Rabbit Polyclonal Antibody

Rabbit polyclonal antibody raised against partial synthetic peptide of human VIM.

  • High specificity
  • High reactivity

Specifications

Antigen VIM
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial synthetic peptide of human VIM.
Dilution ELISA (0.05-0.2 ug/mL) Immunohistochemistry (5-10 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 0.5 mg/mL PBS, pH 7.4 (0.02% sodium azide).
Gene Accession No. P08670
Gene Alias FLJ36605
Gene Symbols VIM
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to residues within 300-350 of human VIM.
Purification Method Immunoaffinity purification
Quantity 40 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7431
Target Species Human
Content And Storage Store at -20°C or lower.
Reconstitute the lyophilized powder with deionized water (or equivalent) to an antibody concentration of 0.5 mg/mL.
The reconstituted antibody can be stored for 2-3 weeks at 4°C. For long term storage, aliquot and st
Form Lyophilized
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.