Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vimentin Antibody (211), DyLight 350

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289211UV
Description
Vimentin Monoclonal specifically detects Vimentin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Vimentin | |
Monoclonal | |
DyLight 350 | |
FLJ36605, vimentin | |
This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the Human Vimentin. | |
0.1 mL | |
Biologically Active Proteins, Cancer, Cell Biology, Cellular Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Markers, Stem Cells | |
7431 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
211 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction