Learn More
Vimentin Antibody (280618), Alexa Fluor™ 594, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB2105T
Description
Vimentin Monoclonal antibody specifically detects Vimentin in Human, Mouse, Rat samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry, KnockDownSpecifications
Vimentin | |
Monoclonal | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry | |
280618 | |
Western Blot, Simple Western, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, Knockout Validated | |
epididymis secretory sperm binding protein, FLJ36605, VIM, vimentin | |
E. coli-derived recombinant human Vimentin, Ser2-Glu466, Accession # P08670 | |
0.1 mL | |
Cancer, Cell Biology, Cellular Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Stem Cells | |
7431 | |
Store at 4C in the dark. | |
IgG2a |