Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vimentin Antibody (280618), DyLight 680, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB2105Y
Description
Vimentin Monoclonal antibody specifically detects Vimentin in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Gene Knock-OutSpecifications
Vimentin | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Simple Western, Immunohistochemistry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Gene Knock-Out | |
280618 | |
Western Blot, Simple Western, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, Knockout Validated | |
epididymis secretory sperm binding protein, FLJ36605, VIM, vimentin | |
E. coli-derived recombinant human Vimentin, Ser2-Glu466, Accession # P08670 | |
0.1 mL | |
Cancer, Cell Biology, Cellular Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Stem Cells | |
7431 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction