Learn More
Vimentin Antibody (2A52), DyLight 650, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP192688C
Catalog No. NBP192688C
Only null left
Add to Cart
Description
Description
Vimentin Monoclonal specifically detects Vimentin in Human, Rat, Mouse (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated.
Specifications
Specifications
Vimentin
Monoclonal
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
VIM
Full length recombinant human vimentin protein expressed in and purified from E. coli.
0.1 mL
Biologically Active Proteins, Cancer, Cellular Markers, Cytoskeleton Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Markers, Stem Cells
7431
Store at 4C in the dark.
IgG1
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoassay
2A52
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated
FLJ36605, vimentin
Mouse
Protein G purified
RUO
Primary
Human, Rat, Mouse (Negative)
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.