Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vimentin Antibody (2A52), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP192688F
Description
Vimentin Monoclonal specifically detects Vimentin in Human, Rat, Mouse (Negative) samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated.Specifications
Vimentin | |
Monoclonal | |
1mg/mL | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated | |
FLJ36605, vimentin | |
Mouse | |
50 kDa | |
0.1 mL | |
Cancer, Cellular Markers, Cytoskeleton Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Markers, Stem Cells | |
7431 | |
Human, Rat, Mouse (Negative) | |
Purified |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoassay | |
2A52 | |
FITC | |
P08670 | |
VIM | |
Full length recombinant human vimentin protein expressed in and purified from E. coli. | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Cow and Porcine. Based upon sequence similarity, this antibody is predicted to react with Chimpanzee (99%), Monkey (99%) and Hamster (97%). Sequence homology of Human Vimentin to Chicken and Xenopus is 86% and 78% respectively. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction