Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vimentin Antibody (2D1), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 5 publications
$244.50 - $499.50
Specifications
Antigen | Vimentin |
---|---|
Clone | 2D1 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:10000, Simple Western 1:100, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin, Knockout Validated |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP192687SS
![]() |
Novus Biologicals
NBP192687SS |
0.025 mL |
Each for $244.50
|
|
|||||
NBP192687
![]() |
Novus Biologicals
NBP192687 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
Vimentin Monoclonal specifically detects Vimentin in Human, Rat, Mouse (Negative) samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
Vimentin | |
1 mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cancer, Cellular Markers, Cytoskeleton Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Stem Cells | |
50% PBS, 50% glycerol with 5mM Sodium Azide | |
FLJ36605, vimentin | |
VIM | |
IgG2a | |
Protein G purified | |
Human, Rat and Mouse and likely all other Mammals. |
2D1 | |
Western Blot 1:10000, Simple Western 1:100, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin, Knockout Validated | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat, Mouse (Negative) | |
P08670 | |
7431 | |
Full length recombinant human Vimentin expressed in and purified from E. coli. [UniProt# P08670] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
50 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title