Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vimentin Antibody (RV203), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197671MFV610
Description
Vimentin Monoclonal antibody specifically detects Vimentin in Human, Mouse, Rat, Porcine, Canine, Goat, Hamster samples. It is validated for Flow CytometrySpecifications
Vimentin | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig, Canine, Goat, Hamster | |
Purified |
Flow Cytometry | |
RV203 | |
Flow Cytometry | |
epididymis secretory sperm binding protein, FLJ36605, VIM, vimentin | |
This antibody is a mouse monoclonal IgG1 antibody derived by fusion of SP2/0-Ag14 Mouse myeloma cells with spleen cells from a BALB/c Mouse immunized with a cytoskeletal vimentin extract of calf lens. | |
0.1 mL | |
Cancer, Cell Biology, Cellular Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Stem Cells | |
7431 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction