Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vimentin Antibody (VIM/1937R), Alexa Fluor™ 532, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308936AF532
Description
Vimentin Monoclonal antibody specifically detects Vimentin in Human, Rat, Porcine, Bovine, Canine, Chicken, Equine, Feline, Mouse (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Vimentin | |
Monoclonal | |
Alexa Fluor 532 | |
epididymis secretory sperm binding protein, FLJ36605, VIM, vimentin | |
Recombinant full-length human Vimentin protein (Uniprot: P08670) | |
100 μg | |
Cancer, Cell Biology, Cellular Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Stem Cells | |
7431 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
VIM/1937R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Rat, Pig, Bovine, Canine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction