Learn More
Vimentin Mouse anti-Canine, Chicken, Goat, Hamster, Human, Monkey, Mouse, Non-human Primate, Porcine, Rat, Xenopus laevis, Zebrafish, FITC, Clone: RV202, MUbio

Mouse Monoclonal Antibody
Supplier: Vector Laboratories MUB1900L1

Description
The antibody may be stored at 4 C. For prolonged storage prepare appropriate aliquots and store at or below -20 C. Prior to use, an aliquot is thawed slowly in the dark at ambient temperature, spun down again and used to prepare working dilutions by adding sterile phosphate buffered saline (PBS, pH 7.2). Repeated thawing and freezing should be avoided. Working dilutions should be stored at 4 C, not refrozen, and preferably used the same day. If a slight precipitation occurs upon storage, this should be removed by centrifugation. It will not affect the performance or the concentration of the product.
Vimentin Monoclonal antibody specifically detects Vimentin in Canine, Chicken, Goat, Hamster, Human, Monkey, Mouse, Non-human Primate, Porcine, Rat, Xenopus laevis, Zebrafish samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen)Specifications
Vimentin | |
Monoclonal | |
0.1 mL/Test | |
PBS with 0.1% BSA and 0.09% sodium azide; pH | |
P02543, P08670, P09654, P20152, P31000, P48670 | |
Mouse | |
Protein A/G | |
RUO | |
100522394, 100689099, 102182515, 140599, 22352, 420519, 477991, 7431, 81818 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen) | |
RV202 | |
FITC | |
Vim | |
cb28; CTRCT30; epididymis luminal protein 113; FLJ36605; HEL113; intermediate filament; intermediate filament protein; RP11-124N14.1; similar to vimentin; Vim; VIME; Vimentin | |
Vimentin | |
1 mL | |
Primary | |
Canine, Chicken, Goat, Hamster, Human, Monkey, Mouse, Non-human Primate, Pig, Rat, X. laevis, Zebrafish | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.