Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vimentin Antibody (RV202), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197672R
Description
Vimentin Monoclonal specifically detects Vimentin in Human, Mouse, Rat, Porcine, Canine, Chicken, Goat, Hamster, Primate, Xenopus, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Vimentin | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
VIM | |
Clone RV202 is a mouse monoclonal IgG1 antibody derived by fusion of SP2/0-Ag14 mouse myeloma cells with spleen cells from a BALB/c mouse immunized with a cytoskeletal vimentin extract of calf lens. | |
0.1 mL | |
Primary | |
RV202 reacts exclusively with vimentin, which is expressed in mesenchymal cells and mesenchymal derived tumors e.g. lymphoma, sarcoma and melanoma. | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
RV202 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
FLJ36605, vimentin | |
Mouse | |
Protein A or G purified | |
Biologically Active Proteins, Cancer, Cellular Markers, Cytoskeleton Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Markers, Stem Cells | |
7431 | |
Human, Mouse, Rat, Pig, Canine, Chicken, Goat, Hamster, Primate, Xenopus species, Zebrafish | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction