Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vimentin, p Ser39 Antibody (SR1953), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322613100UL
Description
Vimentin Monoclonal antibody specifically detects Vimentin in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Vimentin | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
SR1953 | |
Western Blot 1:500-1:2000 | |
epididymis secretory sperm binding protein, FLJ36605, VIM, vimentin | |
A synthesized peptide derived from human phospho-Vimentin [Ser39] (Uniprot #: P08670) | |
100 μg | |
Cancer, Cell Biology, Cellular Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Stem Cells | |
7431 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction