Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vimentin Rabbit anti-Human, Rat, Porcine, Bovine, Canine, Chicken, Equine, Feline, Clone: VIM/1937R, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$316.50

Specifications

Antigen Vimentin
Clone VIM/1937R
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121856
SDP
View Documents
Novus Biologicals
NBP30719120UG
20 μg
Each of 1 for $316.50
Only null left
Add to Cart
 
Description

Description

Vimentin Monoclonal specifically detects Vimentin in Human, Rat, Porcine, Bovine, Canine, Chicken, Equine, Feline samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

Vimentin
0.2 mg/ml
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cancer, Cell Biology, Cellular Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Stem Cells
10 mM PBS with 0.05% BSA
7431
IgG
Protein A or G purified
VIM/1937R
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin
Monoclonal
Purified
RUO
Human, Rat, Pig, Bovine, Canine, Chicken, Equine, Feline
epididymis secretory sperm binding protein, FLJ36605, VIM, vimentin
Recombinant full-length human Vimentin protein
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.