Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vincristine Sulfate, 97+%, PROMO, Thermo Scientific Chemicals

Catalog No. J60907MAPM
Click to view available options
Quantity:
10 mg
25 mg

An anti-tumor alkaloid used in cancer research

Solubility

Soluble in DMSO at 60 mg/mL. Soluble in water at 25 mg/mL with warming. Very poorly soluble in ethanol

Notes

Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away fom strong oxidizing agents, heavy metal salts.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2068-78-2
Molecular Formula C46H58N4O14S
Molecular Weight (g/mol) 923.04 g/mol
MDL Number MFCD00084729
InChI Key AQTQHPDCURKLKT-DJOZZVTQSA-N
PubChem CID 126963557
SMILES CCC1(CC2CC(C3=C(CCN(C2)C1)C4=CC=CC=C4N3)(C5=C(C=C6C(=C5)C78CCN9C7C(C=CC9)(C(C(C8N6C=O)(C(=O)OC)O)OC(=O)C)CC)OC)C(=O)OC)O.OS(=O)(=O)O

Specifications

Type Vincristine Sulfate
Physical Form Powder
Melting Point >300°C
Color White
Quantity 10 mg
UN Number UN2811
Beilstein 3924631
Boiling Point 273°C to 281°C
Merck Index 14,9986
Solubility Information Soluble in DMSO; Soluble in water with warming; Very poorly soluble in ethanol
Percent Purity ≥98%
Chemical Name or Material Vincristine sulfate
Show More Show Less
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 218-190-0
RTECSNumber OH6340000
TSCA No
Recommended Storage Store at -20°C

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.