Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinculin Antibody (VCL/2573), Biotin, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP279941B

Catalog No. NB013056


Only null left
Add to Cart

Description

Description

Vinculin Monoclonal specifically detects Vinculin in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Vinculin
Monoclonal
Biotin
PBS
VCL
Recombinant fragment (around aa 174-322) of human Vinculin (VCL) protein (exact sequence is proprietary) (Uniprot: P18206)
0.1 mL
Cell Biology, Cellular Markers, Phospho Specific
7414
Store at 4°C in the dark.
IgG1 κ
Immunohistochemistry, Immunohistochemistry (Paraffin)
VCL/2573
Immunohistochemistry, Immunohistochemistry-Paraffin
CMD1W, CMH15, Metavinculin, MVCL, vinculin
Mouse
Protein A or G purified
RUO
Primary
Human, Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.