Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vinculin Antibody (VCL/2573), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279941R
Description
Vinculin Monoclonal specifically detects Vinculin in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Vinculin | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
VCL | |
Recombinant fragment (around aa 174-322) of human Vinculin (VCL) protein (exact sequence is proprietary) (Uniprot: P18206) | |
0.1 mL | |
Cell Biology, Cellular Markers, Phospho Specific | |
7414 | |
Store at 4°C in the dark. | |
IgG1 κ |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
VCL/2573 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
CMD1W, CMH15, Metavinculin, MVCL, vinculin | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction