Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Vinculin Antibody (VCL/7091R), Alexa Fluor™ 700, Novus Biologicals™
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP314197AF700
                        
                            
                        
                        
                        
                            
                        
                        
                        
                        
                            
                                    
                                This item is not returnable.
                                View return policy
                                
                        
                    
                Description
Vinculin Monoclonal antibody specifically detects Vinculin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Vinculin | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG | 
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| VCL/7091R | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CMD1W, CMH15, Metavinculin, MVCL, vinculin | |
| Recombinant fragment (around aa 174-322) of human Vinculin (VCL) protein (exact sequence is proprietary) (Uniprot: P18206) | |
| 0.1 mL | |
| Cell Biology, Cellular Markers | |
| 7414 | |
| Store at 4°C in the dark. | |
| Purified | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction