Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vinculin Antibody (VCL/8533R), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32030520UG
Description
Vinculin Monoclonal antibody specifically detects Vinculin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
Vinculin | |
Monoclonal | |
0.2 mg/ml | |
Western Blot 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL | |
CMD1W, CMH15, Metavinculin, MVCL, vinculin | |
Recombinant fragment (around aa 174-322) of human Vinculin protein (exact sequence is proprietary) | |
20 μg | |
Cell Biology, Cellular Markers, Phospho Specific | |
7414 | |
Store at 4&drg;C. Do not freeze. | |
IgG κ |
Western Blot, Immunohistochemistry (Paraffin) | |
VCL/8533R | |
Unconjugated | |
10mM PBS with 0.05% BSA | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction